PRIH93ED88BC Princeton BioMolecular Research, Inc.

5-bromo-2-{[(1E)-(2,4-dimethoxyphenyl)methylene]amino}benzoic acid

  • MDL number MFCD00739944
  • Molecular Weight : 364.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart