PRIH93ED88C4 Princeton BioMolecular Research, Inc.

N-(4-{[(5-ethyl-1,3,4-thiadiazol-2-yl)amino]sulfonyl}phenyl)benzamide

  • MDL number MFCD00561565
  • Molecular Weight : 388.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart