PRIH93ED8FF7 Princeton BioMolecular Research, Inc.

1,3-dichloro-5,7-dimethyladamantane

  • MDL number MFCD31808713
  • Molecular Weight : 233.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart