PRIH93ED9114 Princeton BioMolecular Research, Inc.

(3Z)-5-bromo-1H-indole-2,3-dione 3-hydrazone

  • MDL number MFCD00574003
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart