PRIH9445A4C1 Princeton BioMolecular Research, Inc.

C-(2-Furan-2-yl-cyclopropyl)-methylamine

  • MDL number MFCD16300741
  • Cas Number : 1226290-35-2
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart