PRIH9445A4C3 Princeton BioMolecular Research, Inc.

1-[2-(4-Chloro-phenyl)-cyclopropyl]-ethylamine

  • MDL number MFCD11203840
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart