PRIH9445A4D5 Princeton BioMolecular Research, Inc.

1-Methyl-bicyclopropyl-2-ylamine

  • MDL number MFCD16744492
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart