PRIH9445A4D8 Princeton BioMolecular Research, Inc.

3-(7-azabicyclo[2.2.1]heptan-7-yl)propan-1-ol

  • MDL number MFCD19288557
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart