PRIH9445A4EB Princeton BioMolecular Research, Inc.

3-(2,6-Difluoro-phenyl)-1-methyl-propylamine

  • MDL number MFCD19685924
  • Cas Number : 1315373-41-1
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart