PRIH9445A4F4 Princeton BioMolecular Research, Inc.

2-Phenyl-2-pyrrolidin-1-yl-propylamine

  • MDL number MFCD05197433
  • Molecular Weight : 204.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart