PRIH9445A500 Princeton BioMolecular Research, Inc.

Bicyclopropyl-2-yl-isopropyl-amine

  • MDL number MFCD26381430
  • Molecular Weight : 139.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart