PRIH9445A501 Princeton BioMolecular Research, Inc.

Cyclopentane-1,2-diamine

  • MDL number MFCD07778363
  • Cas Number : 3145-88-8
  • Molecular Weight : 100.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart