PRIH9445A503 Princeton BioMolecular Research, Inc.

2'-Phenyl-bicyclopropyl-2-carboxylic acid

  • MDL number MFCD26381474
  • Molecular Weight : 202.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart