PRIH9445A504 Princeton BioMolecular Research, Inc.

3-Methoxy-2-methyl-2-phenyl-cyclopropanecarboxylic acid

  • MDL number MFCD26381491
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart