PRIH9445A539 Princeton BioMolecular Research, Inc.

2-(2-Ethyl-phenyl)-cyclopropanecarboxylic acid

  • MDL number MFCD16746440
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart