PRIH9445A53A Princeton BioMolecular Research, Inc.

3-(3-Amino-propyl)-phenol

  • MDL number MFCD00870506
  • Cas Number : 56522-50-0
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart