PRIH9445A53D Princeton BioMolecular Research, Inc.

2-(5-Bromo-thiophen-2-yl)-cyclopropylamine

  • MDL number MFCD12068293
  • Molecular Weight : 218.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart