PRIH9445A54E Princeton BioMolecular Research, Inc.

4',4'-dimethyl-3',4'-dihydro-2'H-spiro[cyclopropane-1,1'-naphthalene]-2-carboxamide

  • MDL number MFCD30534489
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart