PRIH9445A551 Princeton BioMolecular Research, Inc.

tert-butyl [2-(benzyloxy)cyclohexyl]carbamate

  • MDL number MFCD24448504
  • Cas Number : 945652-32-4
  • Molecular Weight : 305.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart