PRIH9445A55A Princeton BioMolecular Research, Inc.

1-methyl-4-[(2-phenylcyclopropyl)sulfonyl]benzene

  • MDL number MFCD30534494
  • Molecular Weight : 272.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart