PRIH9445A55C Princeton BioMolecular Research, Inc.

cyclobutylmethyl (2E)-3-cyclobutylacrylate

  • MDL number MFCD30534496
  • Molecular Weight : 194.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart