PRIH9445A562 Princeton BioMolecular Research, Inc.

methyl 7-fluoro-1-oxo-3,4-dihydro-1H-isochromene-3-carboxylate

  • MDL number MFCD30534498
  • Molecular Weight : 224.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart