PRIH9445A565 Princeton BioMolecular Research, Inc.

methyl (8-bromo-2,4-dioxo-1,4-dihydroquinazolin-3(2H)-yl)acetate

  • MDL number MFCD30534500
  • Molecular Weight : 313.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart