PRIH9445A583 Princeton BioMolecular Research, Inc.

2-(3-ethoxyphenyl)cyclopropanecarboxylic acid

  • MDL number MFCD11936032
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart