PRIH9445A590 Princeton BioMolecular Research, Inc.

5-(2,3-difluorophenyl)-2-furaldehyde

  • MDL number MFCD11845340
  • Molecular Weight : 208.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart