PRIH9445A5B2 Princeton BioMolecular Research, Inc.

2-(4-bromo-2-chlorophenyl)cyclopropanecarboxylic acid

  • MDL number MFCD27384066
  • Molecular Weight : 275.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart