PRIH9445A5B9 Princeton BioMolecular Research, Inc.

6-isopropyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine

  • MDL number MFCD16853091
  • Molecular Weight : 196.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart