PRIH9445A5E1 Princeton BioMolecular Research, Inc.

2-(2,5-dimethoxy-4-methylphenyl)cyclopropanamine

  • MDL number MFCD18072209
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart