RIE156317060 Rieke Metals, Inc.

3-(4-Cyano-3-fluorophenyl)-1-propene

  • MDL number MFCD09801087
  • Molecular Weight : 161.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-1081a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1081a
Availability details for
Quantity
  • Availability
  • Add to Cart