RIE156317097 Rieke Metals, Inc.

4-(3-carboethoxyphenyl)-2-chloro-1-butene

  • MDL number MFCD01319579
  • Cas Number : 731772-85-3
  • Molecular Weight : 238.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-142b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-142b
Availability details for
Quantity
  • Availability
  • Add to Cart