RIE156317115 Rieke Metals, Inc.

2-chloro-3-(3-fluorophenyl)-1-propene

  • MDL number MFCD07698625
  • Cas Number : 731773-03-8
  • Molecular Weight : 170.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-172a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-172a
Availability details for
Quantity
  • Availability
  • Add to Cart