RIE156317116 Rieke Metals, Inc.

2-chloro-4-(3-fluorophenyl)-1-butene

  • MDL number MFCD07698626
  • Cas Number : 731773-04-9
  • Molecular Weight : 184.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-172b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-172b
Availability details for
Quantity
  • Availability
  • Add to Cart