RIE156317126 Rieke Metals, Inc.

2-chloro-4-(2-methoxyphenyl)-1-butene

  • MDL number MFCD07698639
  • Cas Number : 731773-13-0
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-192b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-192b
Availability details for
Quantity
  • Availability
  • Add to Cart