RIE156317167 Rieke Metals, Inc.

2-Chloro-3-[(2-trifluoromethyl)phenyl]-1-propene

  • MDL number MFCD09801104
  • Molecular Weight : 220.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-302a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-302a
Availability details for
Quantity
  • Availability
  • Add to Cart