RIE156317271 Rieke Metals, Inc.

4-(3,4-Dimethoxyphenyl)-1-butene

  • MDL number MFCD09801191
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-561b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-561b
Availability details for
Quantity
  • Availability
  • Add to Cart