RIE156317370 Rieke Metals, Inc.

4-(3,4-Difluorophenyl)-1-butene

  • MDL number MFCD07775055
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-731b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-731b
Availability details for
Quantity
  • Availability
  • Add to Cart