RIE156317404 Rieke Metals, Inc.

2-Chloro-3-(2,3,5,6-tetramethylphenyl)-1-propene

  • MDL number MFCD09801279
  • Molecular Weight : 208.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-822a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-822a
Availability details for
Quantity
  • Availability
  • Add to Cart