RIE156317417 Rieke Metals, Inc.

2-Bromo-3-(2-cyano-3-fluorophenyl)-1-propene

  • MDL number MFCD09801292
  • Molecular Weight : 240.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-853a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-853a
Availability details for
Quantity
  • Availability
  • Add to Cart