RIE156317424 Rieke Metals, Inc.

2-Chloro-3-(4-chloro-2-methylphenyl)-1-propene

  • MDL number MFCD09801295
  • Molecular Weight : 201.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-872a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-872a
Availability details for
Quantity
  • Availability
  • Add to Cart