RIE156317505 Rieke Metals, Inc.

3-(5-Ethoxycarbonyl-2-furanyl)-2-methyl-1-propene

  • MDL number MFCD09801352
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10013-004a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10013-004a
Availability details for
Quantity
  • Availability
  • Add to Cart