RIE156317608 Rieke Metals, Inc.

Ethyl 3-cyano-4-fluorobenzoylformate

  • MDL number MFCD09801400
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/14061

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14061
Availability details for
Quantity
  • Availability
  • Add to Cart