RIE156317638 Rieke Metals, Inc.

Ethyl 4-methoxy-2-methylbenzoylformate

  • MDL number MFCD09801430
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/14106

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14106
Availability details for
Quantity
  • Availability
  • Add to Cart