RIE156317652 Rieke Metals, Inc.

Ethyl 2,4,5-trimethylbenzoylformate

  • MDL number MFCD09801443
  • Cas Number : 80120-34-9
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/14123

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14123
Availability details for
Quantity
  • Availability
  • Add to Cart