RIE156317700 Rieke Metals, Inc.

2-Chloro-3-(4-chloro-3-methylphenyl)-1-propene

  • MDL number MFCD07775014
  • Molecular Weight : 201.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-1052a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-1052a
Availability details for
Quantity
  • Availability
  • Add to Cart