RIE156317718 Rieke Metals, Inc.

9-chloro-1-nonene

  • MDL number MFCD00671828
  • Cas Number : 872-06-0
  • Molecular Weight : 160.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/10001-001G

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10001-001G
Availability details for
Quantity
  • Availability
  • Add to Cart