RIE156317723 Rieke Metals, Inc.

3-(3-Fluoro-4-methylphenyl)-1-propene

  • MDL number MFCD06201192
  • Cas Number : 842124-26-9
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-991a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-991a
Availability details for
Quantity
  • Availability
  • Add to Cart