RIE156317795 Rieke Metals, Inc.

3-(2,5-Dimethylphenyl)-1-propene

  • MDL number MFCD09801211
  • Cas Number : 42918-26-3
  • Molecular Weight : 146.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-601a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-601a
Availability details for
Quantity
  • Availability
  • Add to Cart