RIE156317797 Rieke Metals, Inc.

3-(1-Naphthyl)-1-propene

  • MDL number MFCD00021581
  • Cas Number : 2489-86-3
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/10021-001A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10021-001A
Availability details for
Quantity
  • Availability
  • Add to Cart