RIE156317798 Rieke Metals, Inc.

Ethyl 5-methyl-5-hexenoate

  • MDL number MFCD00671855
  • Cas Number : 39495-82-4
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/10003-004C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10003-004C
Availability details for
Quantity
  • Availability
  • Add to Cart