RIE156317815 Rieke Metals, Inc.

2-chloro-1-octene

  • MDL number MFCD00671866
  • Cas Number : 31283-43-9
  • Molecular Weight : 146.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10004-002e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10004-002e
Availability details for
Quantity
  • Availability
  • Add to Cart